Order Entry
Canada
ContactUsLinkComponent
Human Recombinant NAPRTase (from E. coli)
Human Recombinant NAPRTase (from E. coli)
Catalog # 75794-320
Supplier:  Prosci
CAS Number:  
Human Recombinant NAPRTase (from E. coli)
Catalog # 75794-320
Supplier:  Prosci
Supplier Number:  90-363
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 μg
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at –80 °C.
  • Enzyme name:
    NAPRTase
  • Enzyme activity:
    ~0.27 U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1umol nicotinic acid mononucleotide (NaMN) per min.
  • Enzyme synonyms:
    Nicotinate Phosphoribosyltransferase, FHA-HIT-interacting Protein, FHIP, EC 2.4.2.11
  • Purity:
    >98% (SDS-PAGE)
  • Formulation:
    Liquid, In 50 mM HEPES/KOH pH 7.5 containing 300 mM potassium chloride and 10 mM DTT.
  • Cat. no.:
    75794-320
  • Supplier No.:
    90-363

Specifications

About this item

Human nicotinate phosphoribosyltransferase (NAPRTase) is localized in the cytoplasm and is involved in biological processes such as NAD biosynthetic and metabolic processes, nicotinamide metabolic process, nicotinate nucleotide salvage, response to oxidative stress and water-soluble vitamin metabolic process

This recombinant proteins is for research use only.

Fusion-Tag: His Tag

It functions by catalyzing the conversion of nicotinic acid (NA) to NA mononucleotide (NaMN) and is essential for NA to increase cellular NAD levels and prevent oxidative stress of the cells. It is a crucial factor in the NAD+ biosynthesis pathway. Catalytic activity: Beta-nicotinate D-ribonucleotide + diphosphate = nicotinate + 5-phospho-alpha-D-ribose 1-diphosphate.