Order Entry
Canada
ContactUsLinkComponent
Dechloromonas aromatica (strain RCB) Recombinant Chlorite dismutase (from E. coli), ProSci
Dechloromonas aromatica (strain RCB) Recombinant Chlorite dismutase (from E. coli), ProSci
Catalog # 75791-882
Supplier:  Prosci
CAS Number:  
Dechloromonas aromatica (strain RCB) Recombinant Chlorite dismutase (from E. coli), ProSci
Catalog # 75791-882
Supplier:  Prosci
Supplier Number:  92-660
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Dechloromonas aromatica (strain RCB)
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Chlorite dismutase
  • Enzyme synonyms:
    Chlorite dismutase, Chlorite O(2)-lyase, Daro_2580, Cld
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    31.3 kD
  • Sequence:
    Met35-Asp282
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of PBS, 0.5 mM EDTA, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. no.:
    75791-882
  • Supplier No.:
    92-660

Specifications

About this item

Chlorite dismutase (Cld) found in prokaryotic organisms, also known as Chlorite O2-lyase, is a b-type heme containing enzyme that catalyzes the reduction of chlorite into chloride plus dioxygen

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

The subunit of chlorite dismutase consists of a heme free N-terminal and a heme b containing C-terminal ferredoxin-like fold with high structural homology to the dye-decolorizing peroxidases (DyPs). The physiological role of Cld in prokaryote has been shown that some microorganisms can use perchlorate or chlorate as terminal electron acceptors for anaerobic respiration thereby producing chlorite that must be detoxified. This enzyme has gained attention because it can be used in the development of bioremediation processes, biosensors, and controlled dioxygen production.