Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Recombinant Phosphoglucomutase-2 (from Cells)

Human Recombinant Phosphoglucomutase-2 (from Cells)

0.05 mg
Catalog # 75791-800
Supplier:   Prosci
Supplier #:  92-619
Human Recombinant Phosphoglucomutase-2 (from Cells)
Catalog # 75791-800
Supplier:  Prosci
CAS Number:  
Human Recombinant Phosphoglucomutase-2 (from Cells)

0.05 mg

CAS Number:  
Price
$1,247.54
Log in for availability
Each (50µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Phosphoglucomutase-2 (PGM2) is a member of PGM family, which catalyzes the inter-conversion of sugar phosphates and participates in anabolic and catabolic reactions

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

When cells are grown in glucose, PGM catalyzes the conversion of glucose-6-phosphate to glucose-1-phosphate an important precursor required for the synthesis of UDP glucose and trehalose. PGM2 catalyzes the conversion of the nucleoside breakdown products ribose-1-phosphate and deoxyribose-1-phosphate to the corresponding 5-phosphopentoses, and it may also catalyze the interconversion of glucose-1-phosphate and glucose-6-phosphate. But this protein has low glucose 1,6-bisphosphate synthase activity.

Documents

Specifications

Specifications

  • Size:
    0.05 mg
  • Enzyme type:
    Recombinant
  • Enzyme synonyms:
    Phosphoglucomutase-2, PGM 2, Glucose phosphomutase 2, Phosphodeoxyribomutase, Phosphopentomutase, PGM2
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Sequence:
    Met1-Asp612
  • Source:
    Cells
  • Molecular weight:
    70.5 kD
  • Cat. No.:
    75791-800
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of 20 mM Tris,200 mM NaCl,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    N-6 His tag
  • Species:
    Human
  • Enzyme name:
    Phosphoglucomutase-2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.