Mouse Recombinant Carbonic Anhydrase 12
Carbonic Anhydrase (CA) XII, also known as Car12 and CA12, is an extracellular enzyme involved in the regulation of the microenvironment acidity and tumor malignant phenotype, was originally identified as a protein overexpressed in some types of cancers
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It has showed that CA XII is induced by hypoxia and oestrogen and expressed at high levels on various types of cancer. The enzyme is directly involved in tumour progression, and its inhibition has an anti-tumour effect. Apart from its role in carcinogenesis, the enzyme contributes to various other diseases like glaucoma and arteriosclerotic plaques, among others. CA XII is therefore regarded as promising target for specific therapies, and may be used as a novel prognostic marker in combination with histologic grade of the tumors.
- Size:0.05 mg
- Cat. No.:75791-688
- Enzyme type:Recombinant
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme synonyms:Carbonic anhydrase 12, Carbonate dehydratase XII, Carbonic anhydrase XII, CA-XII, CA12, CAXII
- Formulation:Lyophilized from a 0.2 µm filtered solution of 20 mM Tris, 150 mM NaCl,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Purity:>95% as determined by reducing SDS-PAGE
- Tag sequence:C-6 His tag
- Sequence:Ala25-Ser301
- Species:Mouse
- Enzyme name:Carbonic Anhydrase 12
- Molecular weight:32.4 kD



