Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Sulfotransferase 1C4 (from E. coli)
Human Recombinant Sulfotransferase 1C4 (from E. coli)
Catalog # 75791-570
Supplier:  Prosci
CAS Number:  
Human Recombinant Sulfotransferase 1C4 (from E. coli)
Catalog # 75791-570
Supplier:  Prosci
Supplier Number:  92-500
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Sulfotransferase 1C4
  • Enzyme synonyms:
    SULT1C4, Sulfotransferase, SULT1C2, hCG_27301, REF: C1003
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    14.1 kD
  • Sequence:
    Met1-Lys102
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of 4mM HCl. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. no.:
    75791-570
  • Supplier No.:
    92-500

Specifications

About this item

Human Sulfotransferase (SULT1C4) is an enzyme that in humans is encoded by the SULT1C4 gene, belongs to the sulfotransferase 1 family

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

SULT1C4 is expressed at high levels in fetal lung and kidney and at low levels in fetal heart, adult kidney, ovary and spinal chord. Sulfotransferase utilizes 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor to catalyze the sulfate conjugation of drugs, xenobiotic compounds, hormones, and neurotransmitters. It shows activity towards p-nitrophenol and N-hydroxy-2-acetylamino-fluorene (N-OH-2AAF). SULT1C4 plays an important role incatalyzing the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds.