Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Acylphosphate phosphohydrolase 2 (from E. coli)
Human Recombinant Acylphosphate phosphohydrolase 2 (from E. coli)
Catalog # 75791-200
Supplier:  Prosci
CAS Number:  
Human Recombinant Acylphosphate phosphohydrolase 2 (from E. coli)
Catalog # 75791-200
Supplier:  Prosci
Supplier Number:  92-307
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-GST tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Acylphosphate phosphohydrolase 2
  • Enzyme synonyms:
    Acylphosphatase-2, ACYP2, Acylphosphatase, muscle type isozyme, Acylphosphate phosphohydrolase 2, ACYP
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    37.43 kD
  • Sequence:
    Ser2-Tyr99
  • Formulation:
    Supplied as a 0.2 µm filtered solution of PBS,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays
  • Cat. no.:
    75791-200
  • Supplier No.:
    92-307

Specifications

About this item

ACYP2, also known as Acylphosphatase-2, Acylphosphatase, muscle type isozyme, Acylphosphate phosphohydrolase 2, is a protein which belongs to the acylphosphatase family.ACYP2 contains one acylphosphatase-like domain

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-GST tag

Muscle acylphosphatase and erythrocyte acylphosphatase have been isolated.ACYP2 plays an important role in hydrolyzing the phosphoenzyme intermediate of different membrane pumps. Recombinant human ACYP2 protein, fused to GST-tag at N-terminus, was expressed in E. coli.