Human Recombinant Sterol O-acyltransferase 2 (from E. coli)
Catalog # 75791-012
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:N-Trx, 6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Sterol O-acyltransferase 2
- Enzyme synonyms:Acetyl-CoA acetyltransferase cytosolic, Acetyl-CoA transferase-like protein, Cytosolic acetoacetyl-CoA thiolase, ACTL, ACAT2, SOAT2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:59.1 kD
- Sequence:Met1-Glu397
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB,150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
- Cat. no.:75791-012
- Supplier No.:92-209
Specifications
About this item
ACAT2 is a cytoplasmic enzyme which belongs to the thiolase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-Trx, 6 His tag
ACAT2 takes part in lipid metabolism, lipoprotein assembly, catalyzing cholesterol esterification in mammalian cells. It is responsible for the synthesis of cholesteryl esters which are part of lipoproteins containing apoB. ACAT2 deficiency contributes to severe mental retardation and hypotonus.