Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Ornithine decarboxylase (from E. coli)
Human Recombinant Ornithine decarboxylase (from E. coli)
Catalog # 75791-010
Supplier:  Prosci
CAS Number:  
Human Recombinant Ornithine decarboxylase (from E. coli)
Catalog # 75791-010
Supplier:  Prosci
Supplier Number:  92-208
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His, N-T7 tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Ornithine decarboxylase
  • Enzyme synonyms:
    Ornithine decarboxylase, ODC1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    53.5 kD
  • Sequence:
    Met1-Val461
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays
  • Cat. no.:
    75791-010
  • Supplier No.:
    92-208

Specifications

About this item

Ornithine decarboxylase (ODC1) is an enzyme which belongs to the Orn/Lys/Arg decarboxylase class-II family

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His, N-T7 tag

ODC1 is the rate-limiting enzyme of the polyamine biosynthesis pathway which catalyzes ornithine to putrescine. The activity level for the enzyme varies in response to growth-promoting stimuli and exhibits a high turnover rate in comparison to other mammalian proteins.