Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Caspase 14 (from E. coli)
Human Recombinant Caspase 14 (from E. coli)
Catalog # 75790-900
Supplier:  Prosci
CAS Number:  
Human Recombinant Caspase 14 (from E. coli)
Catalog # 75790-900
Supplier:  Prosci
Supplier Number:  92-153
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Caspase 14
  • Enzyme synonyms:
    Caspase-14, CASP-14, CASP14
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    28.7 kD
  • Sequence:
    Ser2-Gln242
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of 20 mM PB,150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays
  • Cat. no.:
    75790-900
  • Supplier No.:
    92-153

Specifications

About this item

Caspase 14 (CASP14) is an enzyme that belongs to the peptidase C14A family

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

The Caspase 14 protein is complexed of unprocessed caspase-14 and processed 19 kDa (p19) and 10 kDa (p10) subunits. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes, which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. CASP14 has been shown to be processed and activated by Caspase 8 and Caspase 10 in vitro, and by anti-Fas agonist antibody or TNF-related apoptosis inducing ligand in vivo. The expression and processing of this caspase may be involved in keratinocyte terminal differentiation, which is important for the formation of the skin barrier. It is believed to be a non-apoptotic caspase which is involved in epidermal differentiation, keratinocyte differentiation and cornification. CASP14 probably regulates maturation of the epidermis by proteolytically processing filaggrin.