Human Recombinant Sulfotransferase 1A3 (from E. coli)
Catalog # 75790-876
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Sulfotransferase 1A3
- Enzyme synonyms:Sulfotransferase 1A3/1A4, ST1A3/ST1A4, Aryl Sulfotransferase 1A3/1A4, Catecholamine-Sulfating Phenol Sulfotransferase, HAST3, M-PST, Monoamine-Sulfating Phenol Sulfotransferase, Placental Estrogen Sulfotransferase, Sulfotransferase Monoamine-Preferring, Thermolabile Phenol Sulfotransferase, TL-PST, SULT1A3, STM, SULT1A4
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:35.6 kD
- Sequence:Met1-Leu295
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM Tris,100 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-876
- Supplier No.:92-141
Specifications
About this item
Sulfotransferase 1A3/1A4 (SULT1A3) is 295 amino acids in length and localizes to the cytoplasm
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
It is a member of the Sulfotransferase 1 family. SULT1A3 can be found in the liver, colon, kidney, lung, brain, spleen, small intestine, placenta, and leukocytes. SULT1A3 exists as a homodimer and it catalyzes the sulfation of phenolic monoamines, such as dopamine, norepinephrine and serotonin, and phenolic and catecholic drugs.