Human Recombinant Sulfotransferase 2A1 (from E. coli)
Catalog # 75790-860
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Sulfotransferase 2A1
- Enzyme synonyms:Bile Salt Sulfotransferase, Dehydroepiandrosterone Sulfotransferase, DHEA-ST, Hydroxysteroid Sulfotransferase, HST, ST2, ST2A3, Sulfotransferase 2A1, ST2A1, SULT2A1, STD
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:35.2 kD
- Sequence:Ser2-Glu285
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM Tris, 100 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-860
- Supplier No.:92-133
Specifications
About this item
Bile Salt Sulfotransferase (SULT2A1( is a cytosolic enzyme that belongs to the Sulfotransferase 1 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
SULT2A1 is primarily expressed in the liver and adrenal tissues, and to a lesser extent in the kidney. SULT2A1 utilizes 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor, and it catalyze the sulfonation of steroids and bile acids in the liver and adrenal glands. SULT2A1 may have a role in the inherited adrenal androgen excess.