Human Recombinant Sulfotransferase 1C2 (from E. coli)
75790-858
: Prosci
:
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Sulfotransferase 1C2
- Enzyme synonyms:Sulfotransferase 1C2, ST1C2, Sulfotransferase 1C1, SULT1C#1, humSULTC2, SULT1C2, SULT1C1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:36.3 kD
- Sequence:Met1-Leu296
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM Tris, 0.1 M NaCl, pH 8.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-858
- Supplier No.:92-132
Sulfotransferase 1C2 (SULT1C2) is a cytosolic enzyme member of the Sulfotransferase 1 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Human SULT1C2 is primarily expressed in the adult stomach, kidney and thyroid gland, and in the fetal kidney and liver. SULT1C2 catalyzes the sulfate conjugation of drugs, xenobiotic compounds, hormones, and neurotransmitters. SULT1C2 may be involved in the activation of carcinogenic hyroxylamines. It shows activity towards p-nitrophenol and N-hydroxy-2-acetylamino-fluorene (N-OH-2AAF).