Human Recombinant Cystathionine Gamma-Lyase (from E. coli)
Catalog # 75790-820
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Cystathionine Gamma-Lyase
- Enzyme synonyms:Cystathionine Gamma-Lyase, Cysteine-Protein Sulfhydrase, Gamma-Cystathionase, CTH
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:44.7 kD
- Sequence:Met1-Ser405
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, 20% glycerol,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-820
- Supplier No.:92-113
Specifications
About this item
Cystathionine Gamma-Lyase (CTH) belongs to the trans-sulfuration enzymes family
This recombinant protein can be used for biological assays. For research use only.
CTH exists as a homotetramer and interacts with CALM in a calcium-dependent manner. CTH breaks down cystathionine into cysteine, ammonia and 2-oxobutanoate. CTH catalyzes the last step in the trans-sulfuration pathway from methionine to cysteine and has broad substrate specificity. Defects in CTH will lead to cystathioninuria, which is an autosomal recessive phenotype characterized by abnormal accumulation of plasma cystathionine.