Human Recombinant Aldehyde dehydrogenase 1-A1 (from E. coli)
Catalog # 75790-788
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Aldehyde dehydrogenase 1-A1
- Enzyme synonyms:Retinal Dehydrogenase 1, RALDH 1, RalDH1, ALDH-E1, ALHDII, Aldehyde Dehydrogenase Family 1 Member A1, Aldehyde Dehydrogenase Cytosolic, ALDH1A1, ALDC, ALDH1, PUMB1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:57 kD
- Sequence:Met1-Ser501
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-788
- Supplier No.:92-097
Specifications
About this item
Aldehyde Dehydrogenase Family 1 Member A1 (ALDH1A1) is a cytoplasmic enzyme that belongs to the Aldehyde Dehydrogenase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
ALDH1A1 is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties and subcellular localizations. ALDH1A1 is the main cytosolic isoform, which has a lower affinity for aldehydes than the mitochondrial enzyme. ALDH1A1 binds free retinal and cellular retinol-binding protein-bound retinal. It can convert/oxidize retinaldehyde to retinoic acid.