Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Carbonic Anhydrase 10 (from E. coli)
Human Recombinant Carbonic Anhydrase 10 (from E. coli)
Catalog # 75790-710
Supplier:  Prosci
CAS Number:  
Human Recombinant Carbonic Anhydrase 10 (from E. coli)
Catalog # 75790-710
Supplier:  Prosci
Supplier Number:  92-058
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Carbonic Anhydrase 10
  • Enzyme synonyms:
    Carbonic Anhydrase-Related Protein 10, Carbonic Anhydrase-Related Protein X, CA-RP X, CARP X, Cerebral Protein 15, CA10, Hucep-15, UNQ533/PRO1076
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    34.1 kD
  • Sequence:
    Ala21-Asn300
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of 25 mM Tris, 150 mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-710
  • Supplier No.:
    92-058

Specifications

About this item

Carbonic Anhydrase-Related Protein 10 (CA10) belongs to the Carbonic Anhydrase family of Zinc Metalloenzymes

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

It is an acatalytic member of the alpha-carbonic anhydrase subgroup. CA10 expression is detected in the adult total brain and almost all parts of the central nervous system, but not in the fetal brain. CA10 catalyze the reversible hydration of carbon dioxide in various biological processes, which is fundamental to many processes such as respiration, renal tubular acidification and bone resorption. It is thought to play a role in the central nervous system, especially in brain development.