To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Environmentally Preferable:
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:Red Blood Cells Page Spot 12|KIAA0106|PRDX6|Peroxiredoxin-6|24 kDa Protein|1-Cys PRX|AOP2|Liver 2D PAGE Spot 40|NSGPx|Acidic Calcium-Independent Phospholipase A2|Non-Selenium Glutathione Peroxidase|Antioxidant Protein 2|aiPLA2|1-Cys Peroxiredoxin
- Protein/peptide name:Peroxiredoxin 6
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:27.2 kD
- Sequence:Met1-Pro224
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:used for biological assays
- Cat. no.:75790-584
- Supplier No.:91-974
Specifications
About this item
Peroxiredoxin 6 belongs to the thiol-specific antioxidant protein family, which is a bifunctional enzyme with two distinct active sites. Peroxiredoxin 6 is involved in redox regulation of the cell and can reduce Hydrogen peroxide and short chain organic, fatty acid, and phospholipid hydroperoxides. Peroxiredoxin 6 may regulates phospholipid turnover and protectes against oxidative injury. Peroxiredoxin 6 eases the oxidative stress and TGF-beta-induced abnormalities of human trabecular meshwork cells, it is necessary for blood vessel integrity in injured skin.
- Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.