Human Recombinant Carboxypeptidase M (from Cells)
75790-334
: Prosci
:
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Carboxypeptidase M
- Enzyme synonyms:Carboxypeptidase M, CPM
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:47.3 kD
- Sequence:Leu18-His422
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,1 mM ZnCl₂, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-334
- Supplier No.:91-841
These enzymes remove C-terminal amino acids from peptides and proteins and exert roles in the physiological processes of blood coagulation/fibrinolysis, inflammation, food digestion and pro-hormone and neuropeptide processing
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
CPM is widely distributed in a variety of tissues and cells. CPM is involved in peptide metabolism on both the cell surface and in extracellular fluids. CPM functions not only as a protease but also as a binding partner in cell-surface protein-protein interactions.