Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant PPIase FKBP2 (from Cells)
Catalog # 75790-330
Supplier:  Prosci
CAS Number:  
Human Recombinant PPIase FKBP2 (from Cells)
Catalog # 75790-330
Supplier:  Prosci
Supplier Number:  91-839
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    PPIase FKBP2
  • Enzyme synonyms:
    Peptidyl-prolyl cis-trans isomerase FKBP2(FKBP2 for short), also named 13 kDa FK506-binding protein, FK506-binding protein 2, Immunophilin FKBP13, Rotamase, is a endoplasmic reticulum peripheral membrane protein., FKBP13
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    14.3 kD
  • Sequence:
    Ala22-Leu142
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Used for biological assays
  • Cat. no.:
    75790-330
  • Supplier No.:
    91-839

Specifications

About this item

Peptidyl-prolyl cis-trans isomerase FKBP2(FKBP2 for short), also named 13 kDa FK506-binding protein, FK506-binding protein 2, Immunophilin FKBP13, Rotamase, is a endoplasmic reticulum peripheral membrane protein

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

It contains 1 PPIase FKBP-type domain and belongs to the FKBP-type PPIase family, FKBP2 subfamily which takes part in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP2 is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin. FKBP2 functions as an ER chaperone and as a component of membrane cytoskeletal scaffolds.