Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Protein Disulfide-Isomerase A4 (from Cells)
Human Recombinant Protein Disulfide-Isomerase A4 (from Cells)
Catalog # 75790-176
Supplier:  Prosci
CAS Number:  
Human Recombinant Protein Disulfide-Isomerase A4 (from Cells)
Catalog # 75790-176
Supplier:  Prosci
Supplier Number:  91-762
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Protein Disulfide-Isomerase A4
  • Enzyme synonyms:
    Protein Disulfide-Isomerase A4, Endoplasmic Reticulum Resident Protein 70, ER Protein 70, ERp70, Endoplasmic Reticulum Resident Protein 72, ER Protein 72, ERp-72, ERp72, PDIA4
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    71.7 kD
  • Sequence:
    Val21-Leu645
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-176
  • Supplier No.:
    91-762

Specifications

About this item

Protein Disulfide-Isomerase A4 (PDIA4) is an endoplasmic reticulum luminal protein that belongs to the protein disulfide isomerase family

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

Human PDIA4 is synthesized as a 625 amino acid precursor that contains a 20 amino acid signal sequence, and a 625 amino acid mature chain, including three thioredoxin domains. PDIA4 catalyzes the rearrangement of -S-S- bonds in proteins and is thought to be a deoxycytidine kinase. In addition, PDIA4 serves as a proteases protein disulfide isomerase, phospholipase or an arrangement of these.