Human Recombinant Pancreatic Lipase-Related 2 (from Cells)
Catalog # 75790-168
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Pancreatic Lipase-Related 2
- Enzyme synonyms:Pancreatic Lipase-Related Protein 2, PL-RP2, Galactolipase, PNLIPRP2, PLRP2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:51.16 kD
- Sequence:Lys18-Cys469
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,1 mM GaCl₂, 10% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-168
- Supplier No.:91-758
Specifications
About this item
Pancreatic Lipase-Related Protein 2 (PNLIPRP2) is a secreted protein that belongs to the Lipase family of AB hydrolase superfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
PNLIPRP2 is a lipase with broad substrate specificity that contains one PLAT domain. PNLIPRP2 is expressed mainly in pancreas. PNLIPRP2 can hydrolyze both phospholipids and galactolipids. PNLIPRP2 acts preferentially on monoglycerides, phospholipids and galactolipids. PNLIPRP2 also contributes to milk fat hydrolysis.