Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Ribonuclease Pancreatic (from Cells)
Human Recombinant Ribonuclease Pancreatic (from Cells)
Catalog # 75790-138
Supplier:  Prosci
CAS Number:  
Human Recombinant Ribonuclease Pancreatic (from Cells)
Catalog # 75790-138
Supplier:  Prosci
Supplier Number:  91-739
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Ribonuclease Pancreatic
  • Enzyme synonyms:
    Ribonuclease Pancreatic, HP-Rnase, RIB-1, RNase UpI-1, Ribonuclease 1, RNase 1, Ribonuclease A, RNase A, RNASE1, RIB1, RNS1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    15.6 kD
  • Sequence:
    Lys29-Thr156
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, 10% Glycerol, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-138
  • Supplier No.:
    91-739

Specifications

About this item

Ribonuclease Pancreatic is a secreted enzyme that belongs to the pancreatic ribonuclease family

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

RNASE1 is an endonuclease that cleaves internal phosphodiester RNA bonds on the 3'-side of pyrimidine bases. RNASE1 prefers poly(C) as a substrate and hydrolyzes 2',3'-cyclic nucleotides, with a pH optimum near 8.0. RNASE1 acts on single stranded and double stranded RNA.