Human Recombinant Protamine-2 (from Cells)
Catalog # 75789-966
Supplier: Prosci
CAS Number:
Specifications
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:Ribonucleoside-Diphosphate Reductase Subunit M2|RR2|RRM2|Ribonucleotide Reductase Small Chain|Ribonucleotide Reductase Small Subunit|PRM2
- Protein/peptide name:Protamine-2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:45.9 kD
- Sequence:Met1-Phe389
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75789-966
- Supplier No.:91-653
Specifications
About this item
Ribonucleoside-Diphosphate Reductase Subunit M2 (RRM2) belongs to the ribonucleoside diphosphate reductase small chain family. The reductase of RRM2 catalyzes the formation of deoxyribonucleotides from ribonucleotides. Synthesis of the encoded protein (M2) is regulated in a cell-cycle dependent fashion. RRM2 supplies the precursors essential for DNA synthesis. RRM2 catalyzes the biosynthesis of deoxyribonucleotides from the corresponding ribonucleotides. Phosphorylation on Ser-20 relieves the inhibitory effect on Wnt signaling.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.