Order Entry
Canada
Orders LinkContactUsLinkComponent
Mouse Recombinant PLXDC2

Mouse Recombinant PLXDC2

0.05 mg
Catalog # 75789-924
Supplier:   Prosci
Supplier #:  91-632
Mouse Recombinant PLXDC2
Catalog # 75789-924
Supplier:  Prosci
CAS Number:  
Mouse Recombinant PLXDC2

0.05 mg

CAS Number:  
Price
$1,514.86
Log in for availability
Each (50µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Mouse Plexin domain-containing protein 2(PLXDC2) is a single-pass type I membrane protein. The protein is expressed in tumor endothelium and in vessels of some normal tissues, such as the muscle and lung. PLXDC2 can interact with CTTN, and may play a role in tumor angiogenesis.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Documents

Specifications

Specifications

  • Size:
    0.05 mg
  • Protein/peptide name:
    PLXDC2
  • Protein synonyms:
    Plxdc2|Plexin domain-containing protein 2|Tem7r|Tumor endothelial marker 7-related protein
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Glu31-Ala455
  • Molecular weight:
    48.95 kD
  • Cat. No.:
    75789-924
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    C-6 His tag
  • Tested applications:
    Bioassay
  • Species:
    Mouse

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.