Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Serine Protease Inhibitor Kazal-Type 1 (from Cells)
Human Recombinant Serine Protease Inhibitor Kazal-Type 1 (from Cells)
Catalog # 75789-660
Supplier:  Prosci
CAS Number:  
Human Recombinant Serine Protease Inhibitor Kazal-Type 1 (from Cells)
Catalog # 75789-660
Supplier:  Prosci
Supplier Number:  91-492
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at –20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    TATI|Tumor-Associated Trypsin Inhibitor|SPINK1|Serine Protease Inhibitor Kazal-Type 1|Pancreatic Secretory Trypsin Inhibitor|PSTI
  • Protein/peptide name:
    Serine Protease Inhibitor Kazal-Type 1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    7.28 kD
  • Sequence:
    Asp24-Cys79
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM MES, 150mM NaCl, 2mM CaCl2, 1mM DTT, 0.05% Brij35, 10% Glycerol, pH 6.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays
  • Cat. no.:
    75789-660
  • Supplier No.:
    91-492

Specifications

About this item

Serine Protease Inhibitor Kazal-Type 1 (SPINK1) is a trypsin inhibitor that prevent the trypsin-catalyzed premature activation of zymogens within the pancreas

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

Defects in SPINK1 are a cause of pancreatitis (PCTT). A disease characterized by the presence of calculi in pancreatic ducts. It causes severe abdominal pain attacks. Defects in SPINK1 are the cause of susceptibility to tropical calcific pancreatitis (TCP). Recombinant SPINK1 protein (rSPINK1) stimulated cell proliferation in benign RWPE as well as cancerous prostate cells. The research result indicated that the potential of SPINK1 as an extracellular therapeutic target in prostate cancer. In contrast, knockdown of SPINK1 in 22RV1 cells inhibited cell proliferation, cell invasion, and tumor growth in xenograft assays.