Human Recombinant Arylsulfatase A (from Cells)
Catalog # 75789-238
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Arylsulfatase A
- Enzyme synonyms:Arylsulfatase A, ASA, Cerebroside-Sulfatase, ARSA
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:52.9 kD
- Sequence:Arg19-Ala507
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75789-238
- Supplier No.:91-281
Specifications
About this item
Arylsulfatase A (ARSA) is a lysosomal enzyme that breaks down Cerebroside 3-Sulfate, a major constituent of the myelin sheet, into Cerebroside and Sulfate
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
The ARSA deficiency results in Metachromatic Leukodystrophy (MLD), a lysosomal storage disease in the central and peripheral nervous systems with severe and progressive neurological symptoms. Recombinant Human ARSA corresponds to the ARSA mature peptide and has sulfatase activity.