Human Recombinant Hydroxyacid Oxidase 1 (from E. coli)
75789-194
: Prosci
:
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:N-Trx, 6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Hydroxyacid Oxidase 1
- Enzyme synonyms:Hydroxyacid Oxidase 1, HAOX1, Glycolate Oxidase, GOX, HAO1, GOX1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:58.6 kD
- Sequence:Met1-Ile370
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75789-194
- Supplier No.:91-259
Hydroxyacid Oxidase 1 (HAO1) is an enzyme that belongs to the FMN-Dependent alpha-Hydroxy Acid Dehydrogenase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-Trx, 6 His tag
HAO1 contains 1 FMN Hydroxy Ccid Dehydrogenase domain. HAO1 is expressed primarily in the liver and pancreas. This protein has 2-Hydroxyacid Oxidase activity. Most HAO1 is active on the 2-Carbon substrate Glycolate, but it can also be active on 2-Hydroxy fatty acids, with higher activity towards 2-Hydroxy Palmitate and 2-Hydroxy Octanoate.