Human Recombinant Galactose Mutarotase (from E. coli)
Catalog # 75789-192
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Galactose Mutarotase
- Enzyme synonyms:Aldose 1-Epimerase, Galactose Mutarotase, GALM
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:38.8 kD
- Sequence:Ala2-Ala342
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75789-192
- Supplier No.:91-258
Specifications
About this item
Galactose Mutarotase (GALM) is a cytoplasmic enzyme that belongs to the Aldose Epimerase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
GALM is a Mutarotase that converts alpha-Aldose to the beta -Anomer. GALM is active on D-Glucose, L-Arabinose, D-Xylose, D-Galactose, Maltose and Lactose. GALM may be required for normal Galactose metabolism by maintaining the equilibrium of alpha- and beta - anomers of Galactose.