Order Entry
Canada
ContactUsLinkComponent
Human Recombinant PPC-DC (from E. coli)
Human Recombinant PPC-DC (from E. coli)
Catalog # 75789-180
Supplier:  Prosci
CAS Number:  
Human Recombinant PPC-DC (from E. coli)
Catalog # 75789-180
Supplier:  Prosci
Supplier Number:  91-232
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    PPCDC|Phosphopantothenoylcysteine Decarboxylase|COAC|PPC-DC
  • Protein/peptide name:
    PPC-DC
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    24.6 kD
  • Sequence:
    Met1-Ser204
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 50mM NaCl, 1mM DTT, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-180
  • Supplier No.:
    91-232

Specifications

About this item

Phosphopantothenoylcysteine Decarboxylase (PPC-DC) is an essential enzyme in the biosynthesis ofCoenzyme A and catalyzes the decarboxylation of PPC to Phosphopantetheine. PPC-DC catalyzes the decarboxylation of the Cysteine moiety of 4-Phosphopantothenoylcysteine (PPC) to form 4-Phosphopantetheine (PPantSH), this reaction forms part of the biosynthesis of Coenzyme A. The enzyme is a member of the larger family of Cysteine Decarboxylases including the Lantibiotic-Biosynthesizing enzymes EpiD and MrsD, all of which use a tightly bound Flavin cofactor to oxidize the Thiol moiety of the substrate to a Thioaldehyde.

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.