Human Recombinant PMKASE (from E. coli)
Catalog # 75789-172
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:PMKASE
- Enzyme synonyms:Phosphomevalonate Kinase, PMKase, hPMK, PMVK, PMKI
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:24.2 kD
- Sequence:Met1-Leu192
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 100 mM NaCl, 1 mM DTT, 10% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75789-172
- Supplier No.:91-228
Specifications
About this item
Phosphomevalonate kinase (PMVK) is a cytosolic enzyme
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
PMVK can be highly expressed in the heart,skeletal muscle, liver, pancreas, and kidney; it is expressed at lower levels in the brain, lung, and placenta. Induced by sterol, PMVK takes part in isopentenyl diphosphate biosynthesis through the mevalonate pathway. PMVK catalyzes the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate in the fifth reaction of the cholesterol biosynthetic pathway.