Human Recombinant Thimet Oligopeptidase (from E. coli)
75789-034
: Prosci
:
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Thimet Oligopeptidase
- Enzyme synonyms:Thimet Oligopeptidase, Endopeptidase 24.15, MP78, THOP1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:80 kD
- Sequence:Lys2-Cys689
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 100 mM NaCl, 10% Glycerol, pH 7.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:For Biological assays
- Cat. no.:75789-034
- Supplier No.:91-159
Thimet Oligopeptidase (THOP1) belongs to the peptidase M3 family which includes neurolysin and mitochondrial intermediate peptidase
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
THOP1 is located in Cytoplasm. THOP1 is widely expressed in human tissues and can detected in different subcellular locations. THOP1 is preferential cleavage for bonds with hydrophobic residues at P1, P2 and P3' and a small residue at P1' in substrates of 5 to 15 residues. THOP1 is involved in the metabolism of neuropeptides under 20 amino acid residues and degradation of cytoplasmic peptide. In addition, THOP1 also can degrade the beta-amyloid precursor protein and generate amyloidogenic fragments.