Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Recombinant PP1MG (from <i>E. coli</i>)

Human Recombinant PP1MG (from E. coli)

0.05 mg
Catalog # 75788-996
Supplier:   Prosci
Supplier #:  91-140
Human Recombinant PP1MG (from <i>E. coli</i>)
Catalog # 75788-996
Supplier:  Prosci
CAS Number:  
Human Recombinant PP1MG (from E. coli)

0.05 mg

CAS Number:  
Price
$1,174.16
Log in for availability
Each (50µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Protein Phosphatase 1G (PP1MG) is a cytoplasmic protein that belongs to the PP2C family. PPM1G is widely expressed; it is abundant in testis, skeletal muscle, and heart. PPM1G is a negative regulator of cell stress response pathways. PPM1G is responsible for the dephosphorylation of Pre-mRNA splicing factors, an important factor for the formation of functional spliceosome. PPM1G also plays a role in regulating cell cycle progression.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Documents

Specifications

Specifications

  • Size:
    0.05 mg
  • Protein/peptide name:
    PP1MG
  • Protein synonyms:
    PPM1C|Protein Phosphatase 1G|Protein Phosphatase 2C Isoform Gamma|PPM1G|Protein Phosphatase Magnesium-Dependent 1 Gamma|Protein Phosphatase 1C|PP2C-Gamma
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Met317-Asp546
  • Source:
    E. coli
  • Molecular weight:
    26.15 kD
  • Cat. No.:
    75788-996
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Formulation:
    Supplied as a 0.2 um filtered solution of 25mM TrisHCl, 1mM DTT, 1mM EDTA, 2mM β-ME, 20% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    C-6 His tag
  • Tested applications:
    Bioassay
  • Species:
    Human

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.