ContactUsLinkComponent
Nalgene™ HDPE Tamper Evident Closures, Thermo Scientific
Nalgene™ HDPE Tamper Evident Closures, Thermo Scientific
Catalog # 89400-736
undefined
Nalgene™ HDPE Tamper Evident Closures, Thermo Scientific
Catalog # 89400-736
Supplier Number:  342141-0384
Specifications
  • Cap Size:
    38-430 mm
  • Sterility:
    Sterile
  • Description:
    Tamper-Evident Closure, Bulk-Pack
  • Material:
    HDPE
  • Cat. no.:
    89400-736
  • Cap Color:
    Natural
  • Supplier No.:
    342141-0384
Specifications
About this item
Nalgene™ HDPE Tamper Evident Closures are specially designed for use with PET/PETG square media bottles.
  • Available in 38-430 finish
  • Natural HDPE closures are ADFC (Animal Derived Component-Free)
  • Fits Nalgene PET and PETG Media Bottles with 38-430 neck finish (except 125mL PETG Media Bottles)
  • Leakproof Guarantee when used with Nalgene PET and PETG Media Bottles
  • Easy application and breaking of the tamper evident seal ring
  • Gamma irradiated to provide 106 Sterility Assurance Level (SAL)
  • Flat top surface for possible printing or labeling