VITLAB® Griffin Beakers with Molded Graduations, PMP, BrandTech
Catalog #: 10218-518
Supplier: Brandtech
Supplier #: 61095
Log in for availability
About this item
High-quality beakers made of glass-transparent polymethylpentene.
- Glass-clear polymethyl pentene (PMP)
- Moderate chemical resistance
Beakers feature molded graduations for durability.
Certifications: All beakers meet the requirements of ISO 7056, except for 10218-514, 10218-520, and 10218-524, which are variants in addition to ISO 7056.
Customers Who Bought This Also Bought