Polytron® Homogenizer PT 10-35 GT, Kinematica
Catalog #: 97036-082
Supplier: Kinematica
Small Business Enterprise
Specifications
- Description:Homogenizer
- Cat. no.:97036-082
- Supplier No.:11010103
Specifications
About this item
The PT 10-35 GT with its 1200 watt motor can be used for applications with operating volumes up to 10 liters. More than 30 dispersing generator with different geometries, designs, sizes, materials and coatings are available. The electronic control system continuously compares the preselected rpm speed with the actual speed and adjusts it so that the speed remains constant even as the viscosity increases.
Certifications: CE marked.
Customers Who Bought This Also Bought