59-408 Cut Protection Assembler Sleeves, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
100% DuPont™ Kevlar® interlock fabric is lightweight and breathable with a high level of cut resistance, abrasion resistance and comfort.
- CPPT rating of 698 grams (Level 2)
- Provides moderate heat protection
- 100% Kevlar® knitwrist with thumbslot provides added protection in the palm and wrist areas
- Ambidextrous
- Elasticized cuff
Use in the aerospace, appliance manufacturing, automotive, general industry, metal fabrication, and metalworking industries. Uses include aircraft assembly, automotive assembly and painting, maintenance of heavy equipment, and welding.
Certifications: ANSI Level 2 Cut Protection.
Specifications
- Catalog No:
- 80093-462
- 80093-464
- Garment application:
- Industrial, Laboratory
- Industrial, Laboratory
- Gender:
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Fabric:
- DuPont™ Kevlar® interlock
- DuPont™ Kevlar® interlock