SpecMat-H Anti-Static Mats, Bulk Rolls, 1.53 mm Thickness, ACL
Supplier: ACL Staticide
Certificates
About this item
SpecMat-H Mats are static dissipative, single-layer, ESD-safe thermoplastic vinyl mats made for bench tops and other work surfaces
Mats are 100% recyclable, and made in the USA. They are available in 24", 30", and 36" widths. All bulk roll mats are 50' in length.
They also work as an excellent shelving liner. Lifetime warranty on electrical resistance (10⁸ to 10⁹ ohms).
Certifications: Compliant to ANSI/ESDA S20.20.
Frequently Bought Together