About this item
The Mag-Bind® RxnPure Plus Kit allows rapid and reliable isolation DNA from PCR and other enzymatic reactions with high yield.
- Removes excess primers, primer-dimers, dNTPs and salts
- Does not require centrifugation/filtration steps
- Can be adapted and scaled to most standard liquid handling robots
- Available in 96 or 384 well format
The system combines the proprietary DNA extraction chemistries with the reversible nucleic acid-binding properties of magnetic beads that selectively bind PCR amplicons 100 bp and larger and eliminate excess nucleotides, primers, and nonspecific products such as primer dimers.
This kit is designed for both manual and fully automated protocols and may not require reprogramming of liquid handling instruments. It is also compatible with multiple liquid handlers and magnetic processors.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Amplified/modified DNA
- Sample type:PCR and other enzymatic reactions
- Target molecule:DNA
- Description:Mag-Bind® RxnPure Plus Kit