Order Entry
Canada
ContactUsLinkComponent
Mag-Bind® Ultra-Pure Plasmid DNA Kit, Omega Bio-Tek
Mag-Bind® Ultra-Pure Plasmid DNA Kit, Omega Bio-Tek
Supplier:  Omega Bio-Tek
Certificates
Mag-Bind® Ultra-Pure Plasmid DNA Kit, Omega Bio-Tek
Supplier:  Omega Bio-Tek

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid (endotoxin-free)
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Mag-Bind® Ultra-Pure Plasmid DNA Kit, Endotoxin free

Specifications

Related Information

Frequently Bought Together