Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Native Haptoglobin (from Plasma)

Human Native Haptoglobin (from Plasma)

1 mg
Catalog # 10834-706
Human Native Haptoglobin (from Plasma)
Catalog # 10834-706
Supplier:  
CAS Number:  
Human Native Haptoglobin (from Plasma)
Human Native Haptoglobin (from Plasma)

1 mg

CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

≥95% pure native haptaglobin (mixed type) isolated from human plasma

An acute-phase plasma protein found in human plasma at 1-3 mg per 1 ml. Binds hemoglobin, thus preventing loss of iron through the kidneys. Humans are polymorphic for haptoglobin, with three major phenotypes: Hp 1-1, Hp 2-1, and Hp 2-2. While the phenotypic distribution can vary greatly between ethnicities and geographic location, the Hp 2-1 phenotype is the most prevalent phenotype in humans. Plasma concentrations of haptoglobin are highest in individuals with Hp 1-1, intermediate in Hp 2-1 individuals, and lowest in Hp 2-2 individuals. Hp 1-1 is the most effective at binding hemoglobin, and Hp 2-2 is the least effective. This functional difference may be associated with the frequency and severity of epilepsy attacks, as researchers have found a correlation between recurring seizures and the Hp 2-2 phenotype.

Documents

Specifications

Specifications

  • Size:
    1 mg
  • Cat. No.:
    10834-706
  • Protein/peptide type:
    Native
  • Protein/peptide name:
    Haptoglobin
  • Formulation:
    Lyophilized as a salt-free solid.
  • Protein synonyms:
    Haptoglobin b-Chain
  • Purity:
    >95%
  • Species:
    Human
  • Source:
    Plasma
  • Molecular weight:
    Hp 1-1: 86 kDa

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.