Specifications
- Description:Miriad Rapid HCV/HIV Antibody Test POU+
- Supplier No.:815311006012
Specifications
About this item
Miriad Rapid HCV/HIV Antibody Test employs MedMira Rapid Vertical Flow Technology to deliver the simultaneous diagnosis of Hepatitis C and HIV-1/2
Miriad HCV/HIV features a built-in reagent control line, works with whole blood, serum, and plasma specimens, and requires no additional equipment. Test kits can be stored for 12 months at 2–30°C (35.6–86°F).
In three easy steps users can obtain multiple instant results on a single test cartridge.
HBc/HIV/HCV Antibody Test controls are available as an accessory to this test..