You searched for: Serological Pipettes
Reusable glass or disposable polystyrene serological pipettes aspirate and release milliliter liquid amounts using the blow-out method. The narrow tube construction is transparent and may come with or without calibrated graduations in various scales for accurate meniscus measurements. These volumetric transfer instruments come sterile for aseptic cell culturing procedures. Choose between non-plugged or cotton plug options to prevent overfilling. Color-coded for correct size identification, serological pipettes are available in different lengths and capacities.
VWR® Serological Pipette
Supplier: VWR International
These serological plastic pipettes are designed to use with tissue culture work.
Expand 1 Items
VWR® Disposable Serological Pipettes
Supplier: VWR International
Polystyrene, transparent and graduated.
Expand 43 Items
VWR® Disposable Aspirating Pipettes, Polystyrene, Sterile
Supplier: VWR International
VWR® Disposable Aspirating Pipettes are manufactured from polystyrene and supplied sterile.
Expand 5 Items
VWR® Open End Serological Pipettes
Supplier: VWR International
Wide, open end design is ideal for viscous and high particulate solutions, as well as slurries such as Sephadex.
Expand 8 Items
VWR® Volumetric Pipettes, Reusable, Color Coded, Class A
Supplier: VWR International
Made from ASTM E438 Type 1 Class A borosilicate 33 expansion glass and as per ASTM specification E969.
Expand 18 Items
VWR® Disposable Serological Pipettes, Glass
Supplier: VWR International
VWR® Disposable Serological Pipettes are available in sterile or nonsterile formats, with a variety of packaging options to choose from.
Expand 19 Items
PYREX® Serological Pipettes, Plugged
Supplier: Corning
These sterile, glass pipettes, calibrated to deliver with blow-out; offer long slender tapered tips to make pipetting faster and easier.
Expand 5 Items
Corning® Stripette® Disposable Serological Pipettes, Polystyrene, Sterile, Plugged, Corning
Supplier: Corning
PS, transparent, graduated, sterile.
Expand 29 Items
PYREX® Serological Pipettes, Corning
Supplier: Corning
These sterile disposable 1 ml PYREX® serological pipettes, calibrated (TD) with blow-out, offer long, slender, tapered tips to make pipetting go faster and easier.
Expand 26 Items
Disposable Serological Pipettes, Glass, Sterile, Plugged, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
These pipettes feature a uniform pulled tip and are clean, plugged, and ready for use.
Expand 10 Items
Disposable Plastic Serological Pipettes, Argos Technologies
Supplier: Argos Technologies
Argos brand standard serological pipettes are designed for accurate and rapid fluid dispensing
Expand 29 Items
Disposable Mini Plastic Serological Pipettes, Argos Technologies
Supplier: Argos Technologies
Argos brand mini pipettes were developed for use in confined spaces such as laminar hoods
Expand 3 Items
Disposable Standard-Tip Serological Pipettes, Plastic, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Standard-Tip Serological Pipettes are irradiation sterilized and suitable for tissue culture work.
Expand 10 Items
Nunc™ Serological Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Select size and packaging options to advance your research with complete confidence when using Nunc™ Serological Pipettes
Expand 18 Items
Disposable Serological Pipettes, Glass, Sterile, TD Color-Coded, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.
Expand 4 Items
PYREX® Reusable Serological Pipettes, Glass, Corning
Supplier: Corning
Reusable serological pipettes made of PYREX® glass.
Expand 8 Items
KIMAX® Reusable Serological Pipettes, Glass, Color-Coded, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Wide-tip pipettes with large tip opening provide faster delivery and are ideal for use with viscous liquids, slurries, and suspensions
Expand 4 Items
Serological Pipet, Shortie, Thermo Scientific
Supplier: Thermo Fisher Scientific
Plugged and individually wrapped in paper peel packaging to maintain sterility.
Expand 2 Items
Disposable Serological Pipettes, Plastic, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
These accurate, plastic pipettes are sterile, plugged, and ready for use.
Expand 8 Items
Dye Pipettes, Glass, Color-Coded
Supplier: DWK Life Sciences (KIMBLE)
Calibrated to deliver with blowout.
Expand 4 Items
Disposable Serological Pipettes, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Pipettes are non-plugged and nonsterile, with a uniform pulled tip.
Expand 5 Items
Shorty Disposable Serological Pipettes, Glass, Sterile, Plugged, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Calibrated to deliver with blowout after free flow of liquid has ended
Expand 10 Items
KIMAX® Reusable Serological Pipettes, Glass, Color-Coded, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
These KIMAX-51® borosilicate glass pipettes are intended for bacteriological, microbiological, or serological work. Pipettes must be blown out after free delivery to obtain the total rated capacity. Scale is permanent brown stain fused into the surface of uniform bore tubing without etching. Plain, fire-polished top end has been tooled to accept cotton plugging. Color-coding facilitates sorting and selection. Designed from ASTM Specification E1044, Style II requirements.
Expand 9 Items
Falcon® Individually Wrapped Serological Pipettes, Corning
Supplier: Corning
The pipettes graduation alignment system is a fast and simple way to bring graduations to your line of sight. Polystyrene, disposable pipettes for tissue culture, bacteriological, and research applications.
Expand 15 Items
Falcon® Disposable Serological Pipettes, Polystyrene, Sterile, Plugged, Corning
Supplier: Corning
Color-coded pipettes feature both negative and reverse graduations (except on 1 ml pipet).
Expand 2 Items
SP Wilmad-LabGlass Serological Color Coded Class B Pipettes, SP Industries
Supplier: SP Industries
Calibrated 'to deliver' (TD) at 20°C, these borosilicate glass pipettes have a permanently colored amber scale with the zero line at the top
Expand 2 Items
Falcon® Bulk Packaged Serological Pipettes, Corning
Supplier: Corning
The pipette graduation alignment system is a fast and simple way to bring graduations to your line of sight. Polystyrene, disposable pipettes for tissue culture, bacteriological, and research applications.
Expand 4 Items
Falcon® Disposable Aspirating Pipettes, Polystyrene, Sterile, Corning
Supplier: Corning
PS pipettes, non plugged, non graduated, disposable, sterile.
Expand 2 Items
Wobble-not Low-Insertion Force Serological Pipettes, VistaLab
Supplier: VistaLab
These pipettes feature a unique 2-tier step design at the plug end for less jamming and less dripping.
Expand 15 Items
Eppendorf® Serological Pipettes
Supplier: Eppendorf
Eppendorf® completes its portfolio in liquid handling with a new line of serological pipettes