ContactUsLinkComponent
CDI's Moist Mark Plus Marker, Cancer Diagnostics
CDI's Moist Mark Plus Marker, Cancer Diagnostics
Catalog # 10809-808
undefined
CDI's Moist Mark Plus Marker, Cancer Diagnostics
Catalog # 10809-808
Supplier Number:  MP1000
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    CDI's Moist Mark Plus Marker
  • Environmentally Preferable:
  • Cat. no.:
    10809-808
  • Supplier No.:
    MP1000
Specifications
About this item
CDI's Moist Mark Plus Marker writes smooth, dark and fine on all cassettes and all slides.
  • Resistant to all solvents
  • Use for cytology
  • Dotting reference marker
  • Formulated ink
Resistant to all solvents. Writes smooth, dark and fine on all cassettes and all slides. Use for cytology as a "dotting" reference marker as well. Strictest manufacturing standards. Proprietary, formulated ink.