Corning® Gosselin™ Square Petri Dish, 120×120 mm, Corning
Catalog #: 10799-777
Supplier: Corning
Specifications
- Dimensions:120 x 120 mm
- Description:Gosselin™ Square Petri Dish
- Cat. no.:10799-777
- Packaging:14/Sleeve, 252/Case
- Supplier No.:BP124-05
Specifications
About this item
PS. Petri plates manufactured under aseptic cleanroom conditions.
- Manufactured from optically clear virgin polystyrene
- Sterilized by irradiation to SAL 10⁻³
- Supplied with 4 vents to provide consistent gas exchange
- New design to enhance stacked dish stability
Corning® Gosselin™ Square petri dishes are optically clear, with a design that has been optimized for use on automated platforms.
The square design is used in colony picking and antibiogram applications.
Customers Who Bought This Also Bought