Ammonium Hydroxide Solution, 10%, 100 ml MDB, Hach
Catalog # CA11027-150
Supplier: Hach
CAS Number:
Specifications
- Description:Ammonium Hydroxide Solution, 10%
- Size:100 ml MDB
- Cat. no.:CA11027-150
- Supplier No.:14736-32
Specifications
About this item
Used to adjust pH in determination of metals.