Phosphate Standard Solution, 500 mg/L as PO₄ (NIST), 10 ml, Hach
Catalog # CA11016-608
Supplier: Hach
CAS Number:
Specifications
- Concentration:500 mg/L PO₄
- Cat. no.:CA11016-608
- Supplier No.:1424210
- Parameters:Phosphate as PO43- (NIST)
Specifications
About this item
For accuracy checks in phosphate (phosphorus) determinations.
- Simple accuracy checks
- Minimize waste
- Reliable results