ContactUsLinkComponent
Blumenthal Mini Rongeur, OR Grade, Sklar®
  10198-402
 :  Sklar
undefined
Blumenthal Mini Rongeur, OR Grade, Sklar®
  10198-402
 :  Sklar
 :  67-5987
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Blumenthal Mini Rongeur, Delicate
  • Length:
    12 cm (4¾")
  • Material:
    Stainless steel
  • Curvature:
    Straight
  • Cat. no.:
    10198-402
  • Supplier No.:
    67-5987
Sklar's® Blumenthal Mini Rongeur is a double spring, plier instrument used in a variety of procedures to trim and remove bone.
  • Reusable and latex-free
  • Premium OR Grade
They are often used in difficult to reach dental surgical sites. This rongeur has delicate tips angled at 30 degrees.