ContactUsLinkComponent
Jarcho Tenaculum Forceps, OR Grade, Sklar®
Jarcho Tenaculum Forceps, OR Grade, Sklar®
  10184-364
 :  Sklar
undefined
Jarcho Tenaculum Forceps, OR Grade, Sklar®
  10184-364
 :  Sklar
 :  91-1275
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Jarcho Tenaculum Forceps
  • Length:
    7½"
  • Material:
    Stainless steel
  • Curvature:
    Curved
  • Cat. no.:
    10184-364
  • Supplier No.:
    91-1275
Sklar's® Jarcho Tenaculum Forceps are ratcheted, finger ring forceps with inward facing single hooks used for grasping and moving tissue
These forceps are most commonly used in OB/GYN procedures to grasp and move the cervix. The single hooks provide a deep bite on the cervical tissue that greatly eliminates the possibility of shifting. Both the shaft and the shanks of this instrument are narrow and slightly curved. The overall length is 7-1/2 inches.