ContactUsLinkComponent
PYREX® 105° Angled Tube Adapter with Two-Way 24/40 Standard Taper Joints , Corning
PYREX® 105° Angled Tube Adapter with Two-Way 24/40 Standard Taper Joints , Corning
Catalog # 89091-090
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
PYREX® 105° Angled Tube Adapter with Two-Way 24/40 Standard Taper Joints , Corning
Catalog # 89091-090
Supplier:  Corning
Supplier Number:  8940-24
Product available on GSA Advantage®
   Low Manufacturing Impact
Specifications
  • ST Joint:
    24/40
  • Description:
    PYREX® 105° Angled Tube Adapter with Two-Way [ST] Joints
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Cat. no.:
    89091-090
  • Supplier No.:
    8940-24
Specifications
About this item
These angled two-way adapters are made of PYREX® borosilicate glass.
  • Arms are approximately 105° apart
  • Used as a receiver adapter
  • Very good chemical and temperature resistance
One end is equipped with an outer 24/40 Standard Taper joint and the other end has an inner 24/40 Standard Taper joint. Used as a receiver adapter.