Order Entry
Canada
ContactUsLinkComponent
PYREX® Low Actinic Condenser, with Drip Tip and 24/40 [ST] Inner Joint, Corning
PYREX® Low Actinic Condenser, with Drip Tip and 24/40 [ST] Inner Joint, Corning
Catalog # 23127-022
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
PYREX® Low Actinic Condenser, with Drip Tip and 24/40 [ST] Inner Joint, Corning
Catalog # 23127-022
Supplier:  Corning
Supplier Number:  52300-650
Product available on GSA Advantage®
   Low Manufacturing Impact

Specifications

  • Description:
    PYREX® 650mm Low Actinic Condenser, with Drip Tip and 24/40 [ST] Inner Joint
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Cat. no.:
    23127-022
  • Supplier No.:
    52300-650
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey

Specifications

About this item

This PYREX® air condenser is made from low actinic glass.

  • Air Reflux style condenser
  • Has an inner 24/40 standard taper joint with a drip tip

To protect light sensitive materials and is useful for condensing materials with boiling points above 150 °C (302 °F).