PYREX® Dual Plug Buret, White Enamel Markings, with Rotaflo® Stopcock, Corning
Supplier: Corning
Certificates
About this item
Burets, made of PYREX® borosilicate glass, have an automatic zeroing device and dual Rotaflo® stopcocks for easy controlled filling.
- Greaseless dual stopcocks allow smoother control and help eliminate titrating errors
- With white enamel markings
Specifications
- Catalog No:
- 17545-505
- 17545-549
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey